Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Annexin A13 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF4149SP
This item is not returnable.
View return policy
Description
Annexin A13 Polyclonal specifically detects Annexin A13 in Human, Mouse samples. It is validated for Western Blot.Specifications
Annexin A13 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
annexin A13, Annexin XIII, Annexin-13, ANX13, ANX13annexin, intestine-specific, ANXA13, Intestine-specific annexin, ISA, ISAannexin-13, MGC150460 | |
Goat | |
Affinity Purified | |
RUO | |
312 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
P27216 | |
ANXA13 | |
E. coli-derived recombinant human Annexin A13 Met1-His316 Accession # P27216 | |
25 μg | |
Primary | |
Detects endogenous human and mouse Annexin A13 in Western blots. In Western blots, this antibody did not cross-react with recombinant human Annexin A1, A2, A3, A4, A5, A6, A7, A8, A9, A10, or A11. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction