Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse APLP-1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB3908SP
This item is not returnable.
View return policy
Description
APLP-1 Monoclonal specifically detects APLP-1 in Human, Mouse samples. It is validated for Western Blot.Specifications
APLP-1 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P51693 | |
APLP1 | |
Mouse myeloma cell line NS0-derived recombinant human APLP-1 Gln34-Glu580 Accession # P51693 | |
25 μg | |
Primary | |
Detects human and mouse APLP-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human APLP-2 or recombinant mouse APLP-2 is observed. | |
Human, Mouse | |
Purified |
Western Blot | |
354020 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
amyloid beta (A4) precursor-like protein 1, amyloid precursor-like protein 1, amyloid-like protein 1, APLP1, APLPAPLP-1 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
333 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction