Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Arginase 1/ARG1 Fluorescein-conjugated Antibody, R&D Systems™

Sheep Polyclonal Antibody has been used in 12 publications
Supplier: R&D Systems IC5868F
This item is not returnable.
View return policy
Description
Arginase 1/ARG1/liver Arginase Polyclonal specifically detects Arginase 1/ARG1/liver Arginase in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Arginase 1/ARG1/liver Arginase | |
Polyclonal | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
AI, ARG1, arginase 1, arginase, liver, arginase-1, EC 3.5.3.1, EC:3.5.3.1, Liver Arginase, Liver-type arginase, PGIF, Type I Arginase | |
Sheep | |
Affinity Purified | |
RUO | |
383 | |
Human, Mouse | |
IgG |
Flow Cytometry | |
Fluorescein | |
P05089 | |
ARG1 | |
E. coli-derived recombinant human Arginase 1/ARG1 Met1-Lys322 Accession # P05089 | |
100 Tests | |
Primary | |
Detects human, mouse, and rat Arginase 1/ARG1 in direct ELISAs and Western blots. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction