Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse ATG7 Alexa Fluor 405-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB6608V100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB6608V100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB6608V100 Supplier R&D Systems Supplier No. FAB6608V100UG
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

ATG7 Monoclonal specifically detects ATG7 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen ATG7
Applications Flow Cytometry
Classification Monoclonal
Clone 683906
Conjugate Alexa Fluor 405
Dilution Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells
Formulation Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide.
Gene Accession No. O95352
Gene Alias APG7, APG7 autophagy 7-like (S. cerevisiae), APG7L, APG7-LIKE, ATG12-activating enzyme E1 ATG7, ATG7 autophagy related 7 homolog (S. cerevisiae), Autophagy-related protein 7, DKFZp434N0735, GSA7, hAGP7, ubiquitin activating enzyme E1-like protein, Ubiquitin-activating enzyme E1-like protein, ubiquitin-like modifier-activating enzyme ATG7
Host Species Mouse
Immunogen E. coli-derived recombinant human ATG7 Asn387-Gln570 Accession # O95352
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10533
Test Specificity Detects human ATG7 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human ATG3, 4A, 4B, 5, 10, 12, or 16L1 is observed.
Target Species Human, Mouse
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only (RUO)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.