Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse ATG7 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems FAB6608N100UG
This item is not returnable.
View return policy
Description
ATG7 Monoclonal specifically detects ATG7 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
ATG7 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
APG7, APG7 autophagy 7-like (S. cerevisiae), APG7L, APG7-LIKE, ATG12-activating enzyme E1 ATG7, ATG7 autophagy related 7 homolog (S. cerevisiae), Autophagy-related protein 7, DKFZp434N0735, GSA7, hAGP7, ubiquitin activating enzyme E1-like protein, Ubiquitin-activating enzyme E1-like protein, ubiquitin-like modifier-activating enzyme ATG7 | |
E. coli-derived recombinant human ATG7 Asn387-Gln570 Accession # O95352 | |
100 μg | |
Primary | |
Detects human ATG7 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human ATG3, 4A, 4B, 5, 10, 12, or 16L1 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
IgG1 |
Flow Cytometry | |
683906 | |
Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
O95352 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
10533 | |
Human, Mouse | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction