Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse ATN1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6567
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6567 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6567 Supplier R&D Systems Supplier No. AF6567
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

ATN1 Polyclonal specifically detects ATN1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen ATN1
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL
Gene Accession No. P54259
Gene Alias atrophin 1, atrophin-1, B37, D12S755E, dentatorubral-pallidoluysian atrophy (atrophin-1), Dentatorubral-pallidoluysian atrophy protein, DRPLA, DRPLANOD, HRS, NOD
Gene Symbols ATN1
Host Species Sheep
Immunogen E. coli-derived recombinant human ATN1 Met1-Gln100 Accession # P54259
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1822
Test Specificity Detects human and mouse ATN1 in Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.