Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Bcl-w Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB8241
This item is not returnable.
View return policy
Description
BCL-W/BCL2L2 Monoclonal specifically detects BCL-W/BCL2L2 in Human, Mouse samples. It is validated for Neutralization.Specifications
| BCL-W/BCL2L2 | |
| Monoclonal | |
| LYOPH | |
| Neutralization | |
| Q92843 | |
| BCL2L2 | |
| E. coli-derived recombinant human Bcl-w Met1-Thr172 Accession # Q92843 | |
| 250 μg | |
| Primary | |
| Detects human and mouse Bcl-w. Neutralizes recombinant human and mouse Bcl-w, but not recombinant human Bcl-2 or Bcl-x. | |
| Human, Mouse | |
| Purified |
| Neutralization | |
| 90336 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Apoptosis regulator Bcl-W, Bcl2-L-2, BCL2L2, BCL2-like 2, Bclw, BCL-W, BCLWbcl-2-like protein 2, KIAA0271BCL2-L-2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 599 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction