Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Beclin 1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5295
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5295 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5295 Supplier R&D Systems Supplier No. AF5295
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Produced in sheep immunized with purified, E. coli-derived, recombinant human Beclin 1 (aa 12 - 239; Accession # Q14457). Human Beclin1 specific IgG was purified by affinity chromatography.

Beclin 1 Polyclonal specifically detects Beclin 1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Beclin 1
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL
Gene Accession No. Q14457
Gene Alias ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, BECN1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30
Gene Symbols BECN1
Host Species Sheep
Immunogen E. coli-derived recombinant human Beclin 1 Met12-Gln239 Accession # Q14457
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8678
Test Specificity Detects human and mouse Beclin 1 in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.