Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse beta-1,4-Glucuronyltransferase 1/B4GAT1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6664SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6664SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6664SP Supplier R&D Systems Supplier No. AF6664SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 Polyclonal specifically detects Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O43505
Gene Alias B3GNT1, B3GN-T1, beta-1,3-N-acetylglucosaminyltransferase bGnT-6, BETA3GNTI, EC 2.4.1.149, i-beta-1,3-N-acetylglucosaminyltransferase, iGAT, IGNT, MDDGA13, N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase, Poly-N-acetyllactosamine extension enzyme, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1B3GNT6, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6
Gene Symbols B4GAT1
Host Species Sheep
Immunogen Chinese hamster ovary cell line CHO-derived recombinant human beta-1,4-Glucuronyltransferase 1/B4GAT1 Asp43-Cys415 Accession # O43505
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11041
Test Specificity Detects human and mouse beta-1,3-N-acetylglucosaminyltransferase 1/B3GNT1 in Western blot. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) B4GalT1 and rhB3GNT 2 is observed.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.