Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse beta-1,4-Glucuronyltransferase 1/B4GAT1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6664
This item is not returnable.
View return policy
Description
Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 Polyclonal specifically detects Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 in Human, Mouse samples. It is validated for Western Blot.Specifications
Beta-1,3-N-Acetylglucosaminyltransferase 1/B3GNT1 | |
Polyclonal | |
Unconjugated | |
O43505 | |
B4GAT1 | |
Chinese hamster ovary cell line CHO-derived recombinant human beta-1,4-Glucuronyltransferase 1/B4GAT1 Asp43-Cys415 Accession # O43505 | |
100 μg | |
Primary | |
Detects human and mouse beta-1,3-N-acetylglucosaminyltransferase 1/B3GNT1 in Western blot. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) B4GalT1 and rhB3GNT 2 is observed. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
B3GNT1, B3GN-T1, beta-1,3-N-acetylglucosaminyltransferase bGnT-6, BETA3GNTI, EC 2.4.1.149, i-beta-1,3-N-acetylglucosaminyltransferase, iGAT, IGNT, MDDGA13, N-acetyllactosaminide beta-1,3-N-acetylglucosaminyltransferase, Poly-N-acetyllactosamine extension enzyme, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 1B3GNT6, UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 6 | |
Sheep | |
Affinity Purified | |
RUO | |
11041 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction