Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse BID Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 18 publications
Supplier: R&D Systems AF860
This item is not returnable.
View return policy
Description
BID Polyclonal specifically detects BID in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunoprecipitation.Specifications
BID | |
Polyclonal | |
Unconjugated | |
AAC71064 | |
BID | |
E. coli-derived recombinant mouse BID Met1-Asp195 Accession # AAC71064 | |
100 μg | |
Primary | |
Detects human and mouse BID, a fragment generated in cells treated with anti-Fas and the carboxyl-terminal portion of recombinant BID generated by cleavage with Caspase 8 in Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunoprecipitation 4 ug/10^6 cells | |
apoptic death agonist, BH3 interacting domain death agonist, BH3-interacting domain death agonist, BID isoform ES(1b), BID isoform L(2), BID isoform Si6, desmocollin type 4, FP497, Human BID coding sequence, MGC15319, MGC42355, p22 BID, tbid | |
Goat | |
Affinity Purified | |
RUO | |
637 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction