Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse BMP-8b Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF6305SP
This item is not returnable.
View return policy
Description
BMP-8b Polyclonal specifically detects BMP-8b in Human, Mouse samples. It is validated for Western Blot.Specifications
BMP-8b | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
BMP-8, bone morphogenetic protein 8b, MGC131757, OP-3 | |
Sheep | |
Affinity Purified | |
RUO | |
656 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
NP_031585 | |
BMP8B | |
E. coli-derived recombinant mouse BMP-8b Thr261-His399 Accession # NP_031585 | |
25 μg | |
Primary | |
Detects mouse BMP-8b in direct ELISAs and human and mouse in Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) BMP-8a and less than 1% cross-reactivity with recombinant human (rh) BMP-3b, rhBMP-10, and rm BMP-10 is observed. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction