Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse BMP-8b Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF6305
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF6305 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF6305 Supplier R&D Systems Supplier No. AF6305
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Polyclonal Antibody

BMP-8b Polyclonal specifically detects BMP-8b in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen BMP-8b
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. NP_031585
Gene Alias BMP-8, bone morphogenetic protein 8b, MGC131757, OP-3
Gene Symbols BMP8B
Host Species Sheep
Immunogen E. coli-derived recombinant mouse BMP-8b Thr261-His399 Accession # NP_031585
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 656
Test Specificity Detects mouse BMP-8b in direct ELISAs and human and mouse in Western blots. In direct ELISAs, approximately 50% cross-reactivity with recombinant mouse (rm) BMP-8a and less than 1% cross-reactivity with recombinant human (rh) BMP-3b, rhBMP-10, and rm BMP-10 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.