Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse/Bovine Insulin APC-conjugated Antibody, R&D Systems™

Catalog No. IC1417A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
IC1417A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. IC1417A Supplier R&D Systems Supplier No. IC1417A
This item is not returnable. View return policy

Rat Monoclonal Antibody has been used in 4 publications

Insulin Monoclonal specifically detects Insulin in Human, Mouse, Bovine samples. It is validated for Intracellular Staining by Flow Cytometry.

Specifications

Antigen Insulin
Applications Flow Cytometry
Classification Monoclonal
Clone 182410
Conjugate APC
Dilution Intracellular Staining by Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Gene Alias IDDM2, ILPR, INS, insulin, IRDN, MODY10, proinsulin
Gene Symbols INS
Host Species Rat
Immunogen E. coli-derived recombinant human Insulin
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3630
Test Specificity Detects bovine, human, and mouse insulin.
Target Species Human, Mouse, Bovine
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.