Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Bovine Integrin alpha 6/CD49f Alexa Fluor™ 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB13501G100UG
This item is not returnable.
View return policy
Description
Integrin alpha 6/CD49f Monoclonal specifically detects Integrin alpha 6/CD49f in Human, Mouse, Bovine samples. It is validated for Flow Cytometry.Specifications
Integrin alpha 6/CD49f | |
Monoclonal | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
ITGA6 | |
Mouse mammary tumor cells | |
100 μg | |
3655 | |
Human, Mouse, Bovine | |
Supernatant |
Flow Cytometry | |
GoH3 | |
Flow Cytometry 0.25-1 μg/106 cells | |
CD49 antigen-like family member F, CD49f, CD49f antigen, DKFZp686J01244, FLJ18737, integrin alpha-6, integrin alpha6B, integrin, alpha 6, ITGA6, ITGA6B, Platelet gpl, VLA-6 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human, mouse, and bovine Integrin alpha 6/CD49f. Recognizes an epitope in the extracellular domain of the Integrin alpha 6 subunit. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction