Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Brachyury Antibody, R&D Systems™
GREENER_CHOICESDP

Catalog No. AF2085 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2085 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2085 Supplier Novus Biologicals Supplier No. AF2085
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 94 publications

Brachyury Polyclonal specifically detects Brachyury in Avian∣Chicken∣Cynomolgus Monkey∣Human∣Mouse∣Rat∣Transgenic Mouse∣Xenograft samples. It is validated for ChIP Assay∣ChIP sequencing (ChIP-seq)∣Flow Cytometry∣Flow Cytometry (Intracellular Staining)∣IF/ICC∣Immunocytochemistry∣Immunocytochemistry/Immunofluorescence∣Immunohistochemistry∣Immunohistochemistry (Frozen)∣Immunohistochemistry (Paraffin)∣Immunolocalization∣In vivo Assay∣Western Blot.

Specifications

Antigen Brachyury
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Flow Cytometry, Flow Cytometry (Intracellular Staining), Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunolocalization, In vivo Assay, Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.1-1 ug/mL, Immunohistochemistry 5-15 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL
Gene Accession No. O15178
Gene Alias brachyury protein, MGC104817, Protein T, SAVA, T brachyury (mouse) homolog, T brachyury homolog, T Brachyury Transcription Factor, T, brachyury homolog (mouse), T-Box Transcription Factor T, TBXT, TF, TFT
Gene Symbols T
Host Species Goat
Immunogen E. coli-derived recombinant human Brachyury Ser2-Glu202 Accession # O15178
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6862
Test Specificity Detects human Brachyury in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Avian, Chicken, Cynomolgus Monkey, Human, Mouse, Rat, Transgenic Mouse, Xenograft
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.