Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Brachyury PE-conjugated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Supplier: R&D Systems IC2085P
This item is not returnable.
View return policy
Description
Brachyury Polyclonal specifically detects Brachyury in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Brachyury | |
Polyclonal | |
Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
brachyury protein, MGC104817, Protein T, SAVA, T brachyury (mouse) homolog, T brachyury homolog, T Brachyury Transcription Factor, T, brachyury homolog (mouse), T-Box Transcription Factor T, TBXT, TF, TFT | |
Goat | |
Affinity Purified | |
RUO | |
6862 | |
Human, Mouse | |
IgG |
Flow Cytometry | |
R-PE | |
O15178 | |
T | |
E. coli-derived recombinant human Brachyury Ser2-Glu202 Accession # O15178 | |
100 Tests | |
Primary | |
Detects human Brachyury in direct ELISAs and Western blots. In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) TBX-6 is observed, and approximately 5% cross-reactivity with rhTBX-2, rhTBX-5, and rhTBX-18 is observed. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction