Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse c-Rel Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF2699SP
Change view
Click to view available options
Quantity:
25 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF2699SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF2699SP Supplier R&D Systems Supplier No. AF2699SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody has been used in 2 publications

c-Rel Polyclonal specifically detects c-Rel in Human, Mouse samples. It is validated for Western Blot, Simple Western, Chromatin Immunoprecipitation (ChIP), Immunocytochemistry.

Specifications

Antigen c-Rel
Applications ChIP Assay, Immunocytochemistry
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Chromatin Immunoprecipitation (ChIP) 5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. NP_033070
Gene Alias cRel, C-Rel, C-Rel proto-oncogene protein, I-Rel, oncogene REL, avian reticuloendotheliosis, proto-oncogene c-Rel, REL, v-rel avian reticuloendotheliosis viral oncogene homolog, v-rel reticuloendotheliosis viral oncogene homolog (avian)
Gene Symbols REL
Host Species Goat
Immunogen E. coli-derived recombinant mouse c-Rel Met1-Ile588 Accession # NP_033070
Purification Method Affinity Purified
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5966
Test Specificity Detects human and mouse c-Rel in Western blots.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.