Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Cadherin Pan Specific Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB18385100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB18385100 100 μg
MAB18385SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB18385100 Supplier R&D Systems Supplier No. MAB18385100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

pan Cadherin Monoclonal specifically detects pan Cadherin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen pan Cadherin
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 1006132
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 5-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P12830
Gene Alias Arc-1, CAD1, CAD3, cadherin 1, E-cadherin (epithelial), cadherin 1, type 1, E-cadherin (epithelial), cadherin-1, Cadherin-2, Cadherin-3, calcium-dependent adhesion protein, epithelial, CAM 120/80, CD324, CD324 antigen, CD325, CDH1, CDH2, CDH3, CDHE, CDHP, cell-CAM 120/80, Cell-CAM120/80, ECAD, E-cadherin, ECadherin, Epithelial cadherin, HJMD, LCAM, L-CAM, NCadherin, PCAD, PCadherin, UVOE-Cadherin, uvomorulin
Host Species Mouse
Immunogen Synthetic peptide containing human E-Cadherin, Accession # P12830
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 999
Test Specificity Detects human E-Cadherin, P-Cadherin, N-Cadherin and mouse E-Cadherin in Western blots. Due to the high homology of the immunogen used, this antibody is designed to be pan specific to Cadherins.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.