Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Carbonic Anhydrase VIII/CA8 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB2187SP
This item is not returnable.
View return policy
Description
Carbonic Anhydrase VIII/CA8 Monoclonal specifically detects Carbonic Anhydrase VIII/CA8 in Human, Mouse samples. It is validated for Western Blot.Specifications
Carbonic Anhydrase VIII/CA8 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL | |
P35219 | |
CA8 | |
E. coli-derived recombinant human Carbonic Anhydrase VIII/CA8 Ala2-Gln290 Accession # P35219 | |
25 μg | |
Primary | |
Detects human Carbonic Anhydrase VIII/CA8 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human CA1, 2, 3, 4, 9, 10, 12, 13, or 14 is observed. | |
Human, Mouse | |
Purified |
Western Blot | |
308320 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CA8, CALS, CALScarbonate dehydratase, CAMRQ3, carbonic anhydrase VIIIMGC99509, carbonic anhydrase-like sequence, carbonic anhydrase-related protein, CARP, CARPCA-related protein, CA-VIII, MGC120502 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
767 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction