Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Carbonic Anhydrase XIV/CA14 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2195SP
This item is not returnable.
View return policy
Description
Carbonic Anhydrase XIV/CA14 Polyclonal specifically detects Carbonic Anhydrase XIV/CA14 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carbonic Anhydrase XIV/CA14 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
CA14, Carbonate dehydratase XIV, carbonic anhydrase XIVcarbonic anhydrase 14, carbonic dehydratase, CAXiV, CA-XIV, EC 4.2.1.1 | |
Goat | |
Affinity Purified | |
RUO | |
23632 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
Q9ULX7 | |
CA14 | |
Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase XIV/CA14 Gly19-Met290 Accession # Q9ULX7 | |
25 μg | |
Primary | |
Detects human Carbonic Anhydrase XIV/CA14 in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction