Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Carbonic Anhydrase XIV/CA14 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF2195
This item is not returnable.
View return policy
Description
Carbonic Anhydrase XIV/CA14 Polyclonal specifically detects Carbonic Anhydrase XIV/CA14 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
Carbonic Anhydrase XIV/CA14 | |
Polyclonal | |
Unconjugated | |
Q9ULX7 | |
CA14 | |
Mouse myeloma cell line NS0-derived recombinant human Carbonic Anhydrase XIV/CA14 Gly19-Met290 Accession # Q9ULX7 | |
100 μg | |
Primary | |
Detects human Carbonic Anhydrase XIV/CA14 in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Immunoprecipitation | |
LYOPH | |
Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
CA14, Carbonate dehydratase XIV, carbonic anhydrase XIVcarbonic anhydrase 14, carbonic dehydratase, CAXiV, CA-XIV, EC 4.2.1.1 | |
Goat | |
Affinity Purified | |
RUO | |
23632 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction