Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Carm1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF7277
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF7277 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF7277 Supplier R&D Systems Supplier No. AF7277
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

Carm1 Polyclonal specifically detects Carm1 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry, Knockout Validated.

Specifications

Antigen Carm1
Applications Western Blot, Immunocytochemistry, Immunoassay
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunocytochemistry 5-15 μg/mL, Knockout Validated
Gene Accession No. Q86X55
Gene Alias coactivator-associated arginine methyltransferase 1EC 2.1.1.-, EC 2.1.1, histone-arginine methyltransferase CARM1, PRMT4, PRMT4EC 2.1.1.125, Protein arginine N-methyltransferase 4
Gene Symbols CARM1
Host Species Goat
Immunogen E. coli-derived recombinant human Carm1 Lys209-Leu379 Accession # Q86X55
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10498
Test Specificity Detects human Carm1 in direct ELISAs and Western blots.
Reconstitution Sterile PBS to a final concentration of 0.2 mg/mL.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.