Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Caspase-3 Alexa Fluor 700-conjugated Antibody, R&D Systems™

Rabbit Monoclonal Antibody
Supplier: R&D Systems IC835N100UG
This item is not returnable.
View return policy
Description
Caspase-3 Monoclonal specifically detects Caspase-3 in Human, Mouse samples. It is validated for Intracellular Staining by Flow Cytometry.Specifications
Caspase-3 | |
Monoclonal | |
Alexa Fluor 700 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
KLH-conjugated human Caspase-3 synthetic peptide, CRGTELDCGIETD, Accession No. U26943 | |
100 μg | |
836.0 | |
Human, Mouse | |
Supernatant |
Flow Cytometry | |
269518 | |
Intracellular Staining by Flow Cytometry 0.25-1 μg/106 cells | |
U26943 | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
Primary | |
Detects human and mouse Caspase-3 cleaved at Asp175. No cross-reactivity was detected with the full-length procaspase-3 or other caspases. | |
Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8°C as supplied. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction