Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Caspase-7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB823
This item is not returnable.
View return policy
Description
Caspase-7 Monoclonal specifically detects Caspase-7 in Human, Mouse samples. It is validated for Western Blot, Knockout Validated.Specifications
Caspase-7 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 μg/mL, Knockout Validated | |
P55210 | |
CASP7 | |
E. coli-derived recombinant human Caspase-7 Accession # P55210 | |
100 μg | |
Primary | |
Detects human and mouse precursor Caspase-7 and the large subunit of cleaved Caspase-7. | |
Human, Mouse | |
Purified |
Western Blot, Immunoassay | |
MCH3101.62 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
apoptosis-related cysteine protease, Apoptotic protease Mch-3, CASP7, CASP-7, caspase 7, apoptosis-related cysteine peptidase, caspase-7, Caspase7, EC 3.4.22, EC 3.4.22.60, ICE-like apoptotic protease 3, Lice2 alpha/beta/gamma, Mch3 | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
840 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction