Learn More
Human/Mouse Caspase-8 Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Caspase-8 |
| Applications | Western Blot, KnockDown |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 0.5 ug/mL, Knockout Validated |
| Gene Accession No. | AAC50645 |
| Gene Alias | AIS, androgen receptor, CASP8, Caspase8, DHTRTFM, Dihydrotestosterone receptorHYSP1, HUMARA, Mch5, NR3C4KD, Nuclear receptor subfamily 3 group C member 4, SMAX1SBMA, spinal and bulbar muscular atrophy |
| Gene Symbols | CASP8 |
| Host Species | Goat |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.