Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Cathepsin B Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 3 publications
$200.50 - $655.00
Specifications
Antigen | Cathepsin B |
---|---|
Clone | 173317 |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Classification | Monoclonal |
Description
Cathepsin B Monoclonal specifically detects Cathepsin B in Human, Mouse samples. It is validated for Western Blot.Specifications
Cathepsin B | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
APP secretase, APPS, cathepsin B, Cathepsin B1, CPSBamyloid precursor protein secretase, CTSB, cysteine protease, EC 3.4.22, EC 3.4.22.1 | |
CTSB | |
IgG1 | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
173317 | |
Western Blot | |
Unconjugated | |
Rat | |
Human, Mouse | |
P10605 | |
1508 | |
Mouse myeloma cell line NS0-derived recombinant mouse Cathepsin B His18-Phe339 Accession # P10605 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects the pro and active forms of human and mouse Cathepsin B in direct ELISAs and Western blots. Does not cross-react with recombinant human Cathepsin C, L, L2, O, S, Z, recombinant mouse Cathepsin C, H, L, or Z. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title