Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Cathepsin L Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 9 publications
$200.16 - $2109.88
Specifications
| Antigen | Cathepsin L |
|---|---|
| Clone | 204101 |
| Dilution | Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunohistochemistry, Immunoprecipitation |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB9521SP
![]() |
R&D Systems
MAB9521SP |
25 μg |
Each for $200.16
|
|
|||||
MAB9521100
![]() |
R&D Systems
MAB9521100 |
100 μg |
Each for $567.90
|
|
|||||
MAB9521500
![]() |
R&D Systems
MAB9521500 |
500 μg |
Each for $2,109.88
|
|
|||||
Description
Cathepsin L Monoclonal specifically detects Cathepsin L in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation.Specifications
| Cathepsin L | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL, Immunoprecipitation 25 ug/mL | |
| Monoclonal | |
| Rat | |
| Human, Mouse | |
| P07711 | |
| 1514 | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin L Glu113-Val333 Accession # P07711 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human and mouse Cathepsin L in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human Cathepsin B, C, L2, O, S, or X/Z/P is observed. In Western blots, both the pro and active forms of recombinant human and mouse Cathspsin L are recognized. |
| 204101 | |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cathepsin L, cathepsin L1, CATL, CTSL, CTSL1, CTSLEC 3.4.22.15, EC 3.4.22, FLJ31037, Major excreted protein, MEP | |
| CTSL1 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title