Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse CCR4 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF5207
This item is not returnable.
View return policy
Description
CCR4 Polyclonal specifically detects CCR4 in Human, Mouse samples. It is validated for Western Blot.Specifications
CCR4 | |
Polyclonal | |
Unconjugated | |
P51680 | |
CCR4 | |
E. coli-derived recombinant mouse CCR4 Met1-Glu39, Ala99-Lys111, Ser176-Ile205, Val268-Tyr284 Accession # P51680 | |
100 μg | |
Primary | |
Detects human and mouse CCR4 in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
C-C chemokine receptor type 4, C-C CKR-4, CC-CKR-4HGCN:14099, CCR-4, CD194, CD194 antigen, chemokine (C-C motif) receptor 4, chemokine (C-C) receptor 4, ChemR13, CKR4MGC88293, CMKBR4k5-5, K5-5 | |
Sheep | |
Affinity Purified | |
RUO | |
1233 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction