Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse CDC73 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5508SP
This item is not returnable.
View return policy
Description
CDC73/HRPT2 Polyclonal specifically detects CDC73/HRPT2 in Human, Mouse samples. It is validated for Western Blot.Specifications
CDC73/HRPT2 | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
C1orf28hyperparathyroidism 2 (with jaw tumor), cell division cycle 73, Paf1/RNA polymerase II complex component, homolog (S.cerevisiae), Cell division cycle protein 73 homolog, chromosome 1 open reading frame 28, HPTJT, HPT-JT, HRPT2, HRPT2FLJ23316, Hyperparathyroidism 2 protein, HYX, parafibromin | |
Goat | |
Affinity Purified | |
RUO | |
79577 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 1 ug/mL | |
Q6P1J9 | |
CDC73 | |
E. coli-derived recombinant human CDC73 Met1-Glu260 Accession # Q6P1J9 | |
25 μg | |
Primary | |
Detects human and mouse CDC73 in Western blots. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction