Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse CDK4 Antibody, R&D Systems™

Goat Polyclonal Antibody
Supplier: R&D Systems AF5254
This item is not returnable.
View return policy
Description
CDK4 Polyclonal specifically detects CDK4 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.Specifications
CDK4 | |
Polyclonal | |
Unconjugated | |
P11802 | |
CDK4 | |
E. coli-derived recombinant human CDK4 Met1-Glu303 Accession # P11802 | |
100 μg | |
Primary | |
Detects human and mouse CDK4 in Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Immunocytochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Simple Western 20 ug/mL, Immunocytochemistry 5-15 ug/mL | |
Cell division protein kinase 4, CMM3, cyclin-dependent kinase 4, EC 2.7.11, EC 2.7.11.22, melanoma cutaneous malignant, 3, MGC14458, PSK-J3, PSK-J3cell division kinase 4 | |
Goat | |
Affinity Purified | |
RUO | |
1019 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction