Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse/Chicken RGM-A Antibody, R&D Systems™

Rat Monoclonal Antibody
$200.50 - $514.50
Specifications
Antigen | RGM-A |
---|---|
Clone | 275407 |
Concentration | LYOPH |
Dilution | Western Blot 1 ug/mL |
Applications | Western Blot |
Description
RGM-A Monoclonal specifically detects RGM-A in Human, Mouse, Chicken samples. It is validated for Western Blot.Specifications
RGM-A | |
LYOPH | |
Western Blot | |
Unconjugated | |
Rat | |
Human, Mouse, Chicken | |
Q6PCX7 | |
56963 | |
Mouse myeloma cell line NS0-derived recombinant mouse RGM-A Cys48-Gly421 Accession # Q6PCX7 | |
Primary | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Detects mouse RGM-A in direct ELISAs and Western blots. In direct ELISAs and Western blots, 100% cross-reactivity with recombinant human RGM-A and recombinant chicken RGM is observed. In Western blots, approximately 25% cross-reactivity with recombinant mouse RGM-C is observed. |
275407 | |
Western Blot 1 ug/mL | |
Monoclonal | |
Purified | |
RUO | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
RGM domain family member A, RGM domain family, member A, RGMA, RGMrepulsive guidance molecule A | |
RGMA | |
IgG2a | |
Protein A or G purified from hybridoma culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title