Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse cIAP-2/HIAP-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB817
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB817 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB817 Supplier R&D Systems Supplier No. MAB817
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 5 publications

cIAP-2/HIAP-1 Monoclonal specifically detects cIAP-2/HIAP-1 in Human, Mouse samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.

Specifications

Antigen cIAP-2/HIAP-1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone 315304
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 8-25 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q13489
Gene Alias AIP1, API2, BIRC3, BIRC3 baculoviral IAP repeat containing 3, c-IAP2, CIAP2, HAIP1, HIAP1, HIAP-1, MALT2, MIHC, RNF49
Gene Symbols BIRC3
Host Species Mouse
Immunogen E. coli-derived recombinant human cIAP-2/HIAP-1 Asn2-Ser604 Accession # Q13489
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 330
Test Specificity Detects human and mouse cIAP-2/HIAP-1 in Western blots. Does not cross-react with HEK293 cells transfected with cIAP-1.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.