Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse cIAP Pan-specific Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 12 publications
Supplier: R&D Systems MAB3400SP
This item is not returnable.
View return policy
Description
cIAP (pan) Monoclonal specifically detects cIAP (pan) in Human, Mouse samples. It is validated for Western Blot, Simple Western.Specifications
cIAP (pan) | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Simple Western 10 ug/mL | |
Q13489 | |
E. coli-derived recombinant human cIAP-2 Asn2-Ser604 Accession # Q13489 | |
25 μg | |
Primary | |
Detects human and mouse cIAP-1 and cIAP-2 in Western blots. | |
Human, Mouse | |
Purified |
Western Blot | |
315301 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
330 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction