Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Cited-2 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB5005
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5005 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MAB5005 Supplier R&D Systems Supplier No. MAB5005
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rat Monoclonal Antibody

Cited-2 Monoclonal specifically detects Cited-2 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry.

Specifications

Antigen Cited-2
Applications Western Blot
Classification Monoclonal
Clone 477226
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. O35740
Gene Alias Cbp/p300-interacting transactivator, with Glu/Asp-rich carboxy-terminal domain2,cbp/p300-interacting transactivator 2, Cited2, MRG-1, MRG1melanocyte-specific gene 1-related gene 1, MSG1-related gene 1, MSG-related protein 1, P35srj
Gene Symbols CITED2
Host Species Rat
Immunogen E. coli-derived recombinant mouse Cited-2 Met1-Cys269 Accession # O35740
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10370
Test Specificity Detects human Cited-2 in direct ELISAs and Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.