Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Cleaved Caspase-3 (Asp175) Antibody, R&D Systems™



Rabbit Monoclonal Antibody has been used in 52 publications
Supplier: R&D Systems MAB835SP
This item is not returnable.
View return policy
Description
Caspase-3 Monoclonal specifically detects Caspase-3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Immunocytochemistry.Specifications
Caspase-3 | |
Monoclonal | |
LYOPH | |
Western Blot 0.5 ug/mL, Immunohistochemistry 0.3-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, Immunocytochemistry 8-25 ug/mL | |
U26943 | |
CASP3 | |
KLH-conjugated human Caspase-3 synthetic peptide CRGTELDCGIETD Accession # U26943 | |
25 μg | |
Primary | |
Detects human and mouse Caspase-3 cleaved at Asp175. No cross-reactivity was detected with the full-length procaspase-3 or other caspases. | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry, Flow Cytometry, Immunocytochemistry | |
269518 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Apopain, apoptosis-related cysteine protease, CASP3, CASP-3, caspase 3, apoptosis-related cysteine peptidase, caspase-3, Caspase3, CPP32, CPP-32, CPP32B, CPP32SREBP cleavage activity 1, Cysteine protease CPP32, EC 3.4.22, EC 3.4.22.56, LICE-1, PARP cleavage protease, procaspase3, Protein Yama, SCA-1, Yama | |
Rabbit | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
836 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction