Learn More
Human/Mouse COX-2 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 5 publications
Supplier:  R&D Systems AF4198SP
Catalog No. AF4198SP
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
COX-2 Polyclonal specifically detects COX-2 in Human, Mouse samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, Immunocytochemistry, CyTOF-ready.
Specifications
Specifications
COX-2
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
COX2, COX-2, COX2cyclooxygenase 2b, cyclooxygenase-2, EC 1.14.99, EC 1.14.99.1, GRIPGHS, hCox-2, PGG/HS, PGH synthase 2, PGHS-2, PHS II, PHS-2, PHS-II, prostaglandin G/H synthase 2, prostaglandin G/H synthase and cyclooxygenase, Prostaglandin H2 synthase 2, Prostaglandin-endoperoxide synthase 2, prostaglandin-endoperoxide synthase 2 (prostaglandin G/H synthase andcyclooxygenase), PTGS2
Goat
Affinity Purified
RUO
5743
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Flow Cytometry, Immunocytochemistry, CyTOF
LYOPH
Western Blot 1 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, Immunocytochemistry 5-15 ug/mL, CyTOF-ready
P35354
PTGS2
E. coli-derived recombinant human COX-2 Ala18-Leu604 Accession # P35354
25 μg
Primary
Detects human and mouse COX-2 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human COX-1 is observed.
Human, Mouse
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.
For Research Use Only