Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse CTLA-4 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB386SP
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB386SP 25 μg
MAB386100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. MAB386SP Supplier R&D Systems Supplier No. MAB386SP
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

CTLA-4 Monoclonal specifically detects CTLA-4 in Human, Mouse samples. It is validated for Western Blot, Agonist Activity.

Specifications

Antigen CTLA-4
Applications Western Blot, In vitro Assay
Classification Monoclonal
Clone 1003705
Conjugate Unconjugated
Dilution Western Blot 2 μg/mL, Agonist Activity 0.05-0.4 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. P16410
Gene Alias CD, CD152, CD152 antigen, CD152IDDM12, CD28, celiac disease 3, CELIAC3, CTLA4, CTLA-4, cytotoxic T-lymphocyte antigen 4, cytotoxic T-lymphocyte protein 4, Cytotoxic T-lymphocyte-associated antigen 4, cytotoxic T-lymphocyte-associated protein 4, cytotoxic T-lymphocyte-associated serine esterase-4, GRD4, GSE, ICOS, ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line, CHO-derived human CTLA-4, Ala37-Phe162, Accession # P16410
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 1493
Test Specificity Detects human CTLA-4 in direct ELISAs. Detects human and moue CTLA-4 in Western blots.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.