Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse CX3CR1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 9 publications
Supplier: R&D Systems AF5825
This item is not returnable.
View return policy
Description
CX3CR1 Polyclonal specifically detects CX3CR1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.Specifications
CX3CR1 | |
Polyclonal | |
Unconjugated | |
Q9Z0D9 | |
CX3CR1 | |
E. coli-derived recombinant mouse CX3CR1 Met1-Thr32, Leu92-Lys104, Thr169-Val196, Lys258-Leu274 Accession # Q9Z0D9 | |
100 μg | |
Primary | |
Detects human and mouse CX3CR1 in direct ELISAs and Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
Beta chemokine receptor-like 1, CCRL1, chemokine (C-C) receptor-like 1, chemokine (C-X3-C motif) receptor 1, chemokine (C-X3-C) receptor 1, CMKbRL1, CMK-BRL1, CMK-BRL-1, CMKDR1, CX3C chemokine receptor 1, Fractalkine receptor, G protein-coupled receptor 13, GPR13G-protein coupled receptor 13, GPRV28, V28, V28C-X3-C CKR-1 | |
Goat | |
Affinity Purified | |
RUO | |
1524 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction