Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse CXCL12/SDF-1 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB310100
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB310100 100 μg
MAB310SP 25 μg
MAB310500 500 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB310100 Supplier R&D Systems Supplier No. MAB310100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 38 publications

CXCL12/SDF-1 Monoclonal specifically detects CXCL12/SDF-1 in Human, Mouse samples. It is validated for Neutralization.

Specifications

Antigen CXCL12/SDF-1
Applications Neutralization
Classification Monoclonal
Clone 79014
Conjugate Unconjugated
Dilution Neutralization
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q6ICW0
Gene Alias chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSF, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF1, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1
Gene Symbols CXCL12
Host Species Mouse
Immunogen E. coli-derived recombinant human CXCL12/SDF-1 alpha Lys22-Lys89 Accession # Q6ICW0
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6387
Test Specificity Detects human and mouse CXCL12/SDF-1 alpha and human CXCL12/SDF-1 beta .
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.