Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse CXCL12/SDF-1 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 11 publications
Supplier: R&D Systems BAF310
This item is not returnable.
View return policy
Description
CXCL12/SDF-1 Polyclonal specifically detects CXCL12/SDF-1 in Human, Mouse samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).Specifications
CXCL12/SDF-1 | |
Polyclonal | |
Biotin | |
Q6ICW0 | |
CXCL12 | |
E. coli-derived recombinant human CXCL12/SDF-1 alpha (R&D Systems, Catalog # 350-NS) Lys22-Lys89 Accession # Q6ICW0 | |
50 μg | |
Primary | |
Detects human and mouse CXCL12/SDF-1 in ELISAs and Western blots. In sandwich immunoassays, approximately 20% cross-reactivity with recombinant human (rh) SDF-1 beta is observed and less than 0.1% cross-reactivity with rhIP-10 and recombinant mouse CRG-2 is observed. | |
Human, Mouse | |
IgG |
Western Blot, ELISA | |
LYOPH | |
Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
chemokine (C-X-C motif) ligand 12, C-X-C motif chemokine 12, hIRH, hSDF-1, Intercrine reduced in hepatomas, IRH, PBSF, PBSFSDF-1a, Pre-B cell growth-stimulating factor, SCYB12, SDF1, SDF-1, SDF1A, SDF1B, SDF1SDF-1b, stromal cell-derived factor 1, TLSF, TLSF-a, TLSF-b, TPAR1 | |
Goat | |
Affinity Purified | |
RUO | |
6387 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction