Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse DACT3/Dapper 3 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF5767
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF5767 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. AF5767 Supplier R&D Systems Supplier No. AF5767
Add to Cart
Add to Cart
This item is not returnable. View return policy

Goat Polyclonal Antibody

DACT3/Dapper 3 Polyclonal specifically detects DACT3/Dapper 3 in Human, Mouse samples. It is validated for Western Blot.

Specifications

Antigen DACT3/Dapper 3
Applications Western Blot
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1 ug/mL
Gene Accession No. Q0PHV7
Gene Alias Antagonist of beta-catenin Dapper homolog 3, arginine rich region 1, Arginine-rich region 1 protein, Dapper 3, dapper homolog 3, dapper, antagonist of beta-catenin, homolog 3 (Xenopus laevis), MGC15476, RRR1
Gene Symbols DACT3
Host Species Goat
Immunogen E. coli-derived recombinant mouse DACT3/Dapper 3 Pro486-Lys587 Accession # Q0PHV7
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 147906
Test Specificity Detects human and mouse DACT3/Dapper 3 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse DACT2 is observed.
Reconstitution Reconstitute at 0.2 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.