Learn More
Human/Mouse Desmocollin-1 Alexa Fluor 350-conjugated Antibody, R&D Systems™

Description
Specifications
Specifications
| Antigen | Desmocollin-1 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 772906 |
| Concentration | 0.2 mg/mL |
| Conjugate | Alexa Fluor 350 |
| Dilution | Flow Cytometry 0.25-1 ug/10^6 cells |
| Formulation | Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide |
| Gene Accession No. | P55849 |
| Gene Alias | cadherin family member 1, CDHF1, desmocollin 1, desmocollin-1, Desmocollin1, desmosomal glycoprotein 2/3, DG2/DG3, DSC1 |
| Show More |
For Research Use Only (RUO)
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.