Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Desmocollin-1 Alexa Fluor 488-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Supplier: R&D Systems FAB7367G100UG
This item is not returnable.
View return policy
Description
Desmocollin-1 Monoclonal specifically detects Desmocollin-1 in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
Desmocollin-1 | |
Monoclonal | |
0.2 mg/mL | |
Flow Cytometry 0.25-1 ug/10^6 cells | |
P55849 | |
DSC1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Desmocollin-1 Arg135-Lys691 Accession # P55849 | |
RUO | |
1823 | |
Human, Mouse | |
IgG2a |
Flow Cytometry | |
772906 | |
Alexa Fluor 488 | |
Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
cadherin family member 1, CDHF1, desmocollin 1, desmocollin-1, Desmocollin1, desmosomal glycoprotein 2/3, DG2/DG3, DSC1 | |
Rat | |
100 μg | |
Primary | |
Detects mouse Desmocollin-1 in ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human Desmocollin-1 is observed, and no cross-reactivity with recombinant mouse Desmocollin-2 or -3 is observed. | |
Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction