Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Desmocollin-1 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 2 publications
Supplier: R&D Systems MAB7367
This item is not returnable.
View return policy
Description
Desmocollin-1 Monoclonal specifically detects Desmocollin-1 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF-ready.Specifications
Desmocollin-1 | |
Monoclonal | |
LYOPH | |
Western Blot 0.1 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, Immunohistochemistry 1-25 ug/mL, Immunocytochemistry 8-25 ug/mL, CyTOF-ready | |
P55849 | |
DSC1 | |
Mouse myeloma cell line NS0-derived recombinant mouse Desmocollin-1 Arg135-Lys691 Accession # P55849 | |
100 μg | |
Primary | |
Detects mouse Desmocollin-1 in ELISAs and Western blots. In direct ELISAs, 100% cross-reactivity with recombinant human Desmocollin-1 is observed, and no cross-reactivity with recombinant mouse Desmocollin-2 or -3 is observed. | |
Human, Mouse | |
Purified |
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, CyTOF | |
772906 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
cadherin family member 1, CDHF1, desmocollin 1, desmocollin-1, Desmocollin1, desmosomal glycoprotein 2/3, DG2/DG3, DSC1 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
1823 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction