Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse DGK-epsilon Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7069
This item is not returnable.
View return policy
Description
DGK-epsilon Polyclonal specifically detects DGK-epsilon in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DGK-epsilon | |
Polyclonal | |
Unconjugated | |
P52429 | |
DGKE | |
E. coli-derived recombinant human DGK-epsilon Asn314-Arg435 Accession # P52429 | |
100 μg | |
Primary | |
Detects recombinant human DGK-epsilon in direct ELISAs. Detects human and mouse DGK-epsilon in Western blots. | |
Human, Mouse | |
IgG |
Western Blot, Immunohistochemistry | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
DAG kinase epsilon, DAGK5, DAGK6, DGK, DGKE, DGKepsilon, DGK-epsilon, diacylglycerol kinase epsilon, diacylglycerol kinase, epsilon (64kD), diacylglycerol kinase, epsilon 64kDa, Diglyceride kinase epsilon, EC 2.7.1.107 | |
Sheep | |
Affinity Purified | |
RUO | |
8526 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction