Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse DLL4 Antibody, R&D Systems™

Rat Monoclonal Antibody has been used in 8 publications
Supplier: R&D Systems MAB1389
This item is not returnable.
View return policy
Description
DLL4 Monoclonal specifically detects DLL4 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
DLL4 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunohistochemistry 5-25 ug/mL | |
Q9JI71 | |
DLL4 | |
Mouse myeloma cell line NS0-derived recombinant mouse DLL4 Ser28-Pro525 Accession # Q9JI71 | |
500 μg | |
Primary | |
Detects mouse DLL4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, this antibody shows 100% cross-reactivity with recombinant human DLL4. | |
Human, Mouse | |
Purified |
Western Blot, Immunohistochemistry | |
207822 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
delta 4, Delta 4 precursor, delta ligand 4, delta4, delta-like 4 (Drosophila), delta-like 4 homolog, delta-like 4 homolog (Drosophila), delta-like 4 protein, delta-like protein 4, Drosophila Delta homolog 4, hdelta2, MGC126344, notch ligand delta-2, notch ligand DLL4 | |
Rat | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
54567 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction