Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human/Mouse Doc2 alpha Antibody, R&D Systems™

Sheep Polyclonal Antibody
Supplier: R&D Systems AF7904SP
This item is not returnable.
View return policy
Description
Doc2-alpha Polyclonal specifically detects Doc2-alpha in Human, Mouse samples. It is validated for Western Blot.Specifications
Doc2-alpha | |
Polyclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Doc2, DOC2A, doc2-alpha, double C2-like domain-containing protein alpha, double C2-like domains, alpha | |
Sheep | |
Affinity Purified | |
RUO | |
8448 | |
Sterile PBS to a final concentration of 0.2 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot | |
LYOPH | |
Western Blot 2 ug/mL | |
Q14183 | |
DOC2A | |
E. coli-derived recombinant human Doc2 alpha Met1-Lys114 (Gly48Ser) Accession # Q14183 | |
25 μg | |
Primary | |
Detects human and mouse Doc2 alpha in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human Doc2 beta is observed. | |
Human, Mouse | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction