Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse Doublecortin/DCX Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. AF10025100
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AF10025100 100 μg
AF10025SP 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AF10025100 Supplier R&D Systems Supplier No. AF10025100
Add to Cart
Add to Cart
This item is not returnable. View return policy

Sheep Antibody

Doublecortin specifically detects Doublecortin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Doublecortin
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1 μg/mL, Immunohistochemistry 8-25 μg/mL
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Accession No. O43602
Gene Alias DBCN, Doublecortex, doublecortin, Doublin, FLJ51296, Lissencephalin-X, Lis-X, LISX, X-linked (doublecortin)
Host Species Sheep
Immunogen E.coli-derived recombinant human Doublecortin/DCX, Met116-Thr293, Accession # O43602
Purification Method Antigen Affinity-purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) 1641
Test Specificity Detects human Doublecortin/DCX in direct ELISAs and Western blots. Detects mouse Doublecortin/DCX in immunohistochemistry.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution.
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.