Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Human/Mouse DR3/TNFRSF25 Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. MAB943500
Change view
Click to view available options
Quantity:
100 μg
25 μg
500 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB943500 500 μg
MAB943SP 25 μg
MAB943100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MAB943500 Supplier R&D Systems Supplier No. MAB943500
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody has been used in 2 publications

DR3/TNFRSF25 Monoclonal specifically detects DR3/TNFRSF25 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, CyTOF-ready.

Specifications

Antigen DR3/TNFRSF25
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 59204
Conjugate Unconjugated
Dilution Western Blot 2 ug/mL, Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Gene Accession No. Q93038
Gene Alias Apo-3, APO3, Apoptosis-inducing receptor AIR, Apoptosis-mediating receptor DR3, Apoptosis-mediating receptor TRAMP, Death receptor 3, DR3member 12 (translocatingchain-association membrane protein), LARD, Lymphocyte-associated receptor of death, Protein WSL, Protein WSL-1, TNFRSF25, TRAMP, tumor necrosis factor receptor superfamily, member 25, WSL, WSL1, WSL-1
Gene Symbols TNFRSF25
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human DR3 isoform 1 Gln25-Phe201 Accession # Q93038
Purification Method Protein A or G purified from ascites
Quantity 500 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8718
Test Specificity Detects human DR3/TNFRSF25 in direct ELISAs and Western blots. Shows 100% cross-reactivity with recombinant mouse (rm) DR3/TNFRSF25 and no cross-reactivity with recombinant human (rh) 4-1BB, rhCD27, rhCD30, rhCD40, rhDR6, rhBAFF R, rhFas, rhGITR, rhLTR beta , rhNGF R, rhOPG, rmOX40, rhRANK, rhTAJ, rhTNF RI, rhTNF RII, or rhHVEM.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Mouse
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Ascites
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.